Assay Details
Gene Symbol: LOC4342102 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015788312.2 | XP_015643798.1 | 6 - 7 | 951 | 62 | |
GenBank mRNA | AK073850.1 | - | 7 - 8 | 934 | 62 | |
AK103177.1 | - | 6 - 7 | 595 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
