Assay Details
| Gene Symbol: LOC4342302 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789702.2 | XP_015645188.1 | 1 | 800 | 152 | |
| GenBank mRNA | AK121537.1 | - | 1 | 644 | 152 | |
More Information
Set Membership: |
|

LOC4342302
Genomic Map