Assay Details
| Gene Symbol: LOC4343198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791878.2 | XP_015647364.1 | 1 - 2 | 546 | 144 | |
| GenBank mRNA | AK068617.1 | - | 1 - 2 | 552 | 144 | |
More Information
Set Membership: |
|

LOC4343198
Genomic Map