Assay Details
| Gene Symbol: LOC4343269 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790951.1 | XP_015646437.1 | 1 - 2 | 178 | 89 | |
| GenBank mRNA | AK061330.1 | - | 1 - 2 | 148 | 89 | |
| AK101803.1 | - | 1 - 2 | 149 | 89 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015790951.1 transcripts.
Set Membership: |
|

LOC4343269
Genomic Map