Assay Details
Gene Symbol: LOC4343269 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015790951.1 | XP_015646437.1 | 3 - 4 | 349 | 77 | |
GenBank mRNA | AK061330.1 | - | 3 - 4 | 319 | 77 | |
AK101803.1 | - | 3 - 4 | 320 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
