Assay Details
| Gene Symbol: LOC4343269 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790951.1 | XP_015646437.1 | 4 - 5 | 595 | 114 | |
| GenBank mRNA | AK061330.1 | - | 4 - 5 | 566 | 115 | |
| AK101803.1 | - | 4 - 5 | 566 | 114 | ||
More Information
Set Membership: |
|

LOC4343269
Genomic Map