Assay Details
| Gene Symbol: LOC4343269 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790951.1 | XP_015646437.1 | 5 - 6 | 658 | 84 | |
| GenBank mRNA | AK061330.1 | - | 5 - 6 | 629 | 84 | |
| AK101803.1 | - | 5 - 6 | 629 | 84 | ||
More Information
Set Membership: |
|

LOC4343269
Genomic Map