Assay Details
| Gene Symbol: LOC4343299 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791108.2 | XP_015646594.1 | 3 - 4 | 532 | 88 | |
| XM_015791109.2 | XP_015646595.1 | 3 - 4 | 738 | 88 | ||
| XM_015791110.2 | XP_015646596.1 | 3 - 4 | 475 | 88 | ||
| XM_015791111.2 | XP_015646597.1 | 3 - 4 | 535 | 88 | ||
| XM_015791112.2 | XP_015646598.1 | 3 - 4 | 415 | 88 | ||
| GenBank mRNA | AK103233.1 | - | 3 - 4 | 739 | 88 | |
More Information
Set Membership: |
|

LOC4343299
Genomic Map