Assay Details
| Gene Symbol: LOC4343301 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791591.2 | XP_015647077.1 | 15 - 16 | 2301 | 104 | |
| XM_015791592.2 | XP_015647078.1 | 14 - 15 | 2129 | 104 | ||
| GenBank mRNA | AK068275.1 | - | 15 - 16 | 1761 | 104 | |
More Information
Set Membership: |
|

LOC4343301
Genomic Map