Assay Details
Gene Symbol: LOC4343672 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015789425.2 | XP_015644911.1 | 1 - 2 | 823 | 125 | |
GenBank mRNA | AK111653.1 | - | 1 - 2 | 819 | 125 | |
AK288534.1 | - | 3429 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
