Assay Details
Gene Symbol: LOC4343899 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015792050.1 | XP_015647536.1 | 4 - 5 | 859 | 72 | |
GenBank mRNA | AK065604.1 | - | 4 - 5 | 791 | 72 | |
AK068566.1 | - | 4 - 5 | 789 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
