Assay Details
Gene Symbol: LOC4343903 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015791865.2 | XP_015647351.1 | 2 - 3 | 533 | 72 | |
XM_015791866.2 | XP_015647352.1 | 2 - 3 | 533 | 72 | ||
GenBank mRNA | AK066317.1 | - | 2 - 3 | 457 | 72 | |
AK073803.1 | - | 2 - 3 | 488 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
