Assay Details
Gene Symbol: LOC4343903 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015791866.2 | XP_015647352.1 | 4 - 5 | 2436 | 106 | |
GenBank mRNA | AK066317.1 | - | 4 - 5 | 2360 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
