Assay Details
| Gene Symbol: LOC4344260 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792109.2 | XP_015647595.1 | 4 - 5 | 1111 | 61 | |
| GenBank mRNA | AB183706.1 | - | 4 - 5 | 1092 | 61 | |
| AK099614.1 | - | 4 - 5 | 1231 | 61 | ||
More Information
Set Membership: |
|

LOC4344260
Genomic Map