Assay Details
Gene Symbol: LOC4344526 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015793946.2 | XP_015649432.1 | 2 - 3 | 773 | 109 | |
GenBank mRNA | AK067392.1 | - | 2 - 3 | 780 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
