Assay Details
| Gene Symbol: LOC4344973 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794999.2 | XP_015650485.1 | 1 - 2 | 334 | 79 | |
| XM_015795000.2 | XP_015650486.1 | 1 - 2 | 331 | 79 | ||
| XM_015795001.2 | XP_015650487.1 | 1 - 2 | 315 | 79 | ||
| GenBank mRNA | AK287940.1 | - | 2189 | 79 | ||
More Information
Set Membership: |
|

LOC4344973
Genomic Map