Assay Details
| Gene Symbol: LOC4345095 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792935.2 | XP_015648421.1 | 4 - 5 | 2132 | 78 | |
| XM_015792936.2 | XP_015648422.1 | 4 - 5 | 2136 | 78 | ||
| GenBank mRNA | AK108045.1 | - | 3 - 4 | 637 | 78 | |
More Information
Set Membership: |
|

LOC4345095
Genomic Map