Assay Details
| Gene Symbol: LOC4345386 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792702.2 | XP_015648188.1 | 10 - 11 | 1746 | 99 | |
| XM_015792703.2 | XP_015648189.1 | 10 - 11 | 1731 | 99 | ||
| GenBank mRNA | AK059023.1 | - | 9 - 10 | 1136 | 99 | |
| AK102062.1 | - | 10 - 11 | 1700 | 99 | ||
More Information
Set Membership: |
|

LOC4345386
Genomic Map