Assay Details
| Gene Symbol: LOC4345386 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792702.2 | XP_015648188.1 | 11 - 12 | 1805 | 86 | |
| XM_015792703.2 | XP_015648189.1 | 11 - 12 | 1790 | 86 | ||
| GenBank mRNA | AK059023.1 | - | 10 - 11 | 1195 | 86 | |
| AK102062.1 | - | 11 - 12 | 1759 | 86 | ||
More Information
Set Membership: |
|

LOC4345386
Genomic Map