Assay Details
| Gene Symbol: LOC4345404 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795075.2 | XP_015650561.1 | 8 - 9 | 1890 | 103 | |
| XM_015795076.2 | XP_015650562.1 | 8 - 9 | 1863 | 103 | ||
| XM_015795077.2 | XP_015650563.1 | 8 - 9 | 1853 | 103 | ||
| XM_015795078.2 | XP_015650564.1 | 8 - 9 | 1875 | 103 | ||
More Information
Set Membership: |
|

LOC4345404
Genomic Map