Assay Details
| Gene Symbol: LOC4345926 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792794.2 | XP_015648280.1 | 2 - 3 | 694 | 116 | |
| GenBank mRNA | AK103187.1 | - | 2 - 3 | 658 | 116 | |
More Information
Set Membership: |
|

LOC4345926
Genomic Map