Assay Details
| Gene Symbol: LOC4345926 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792794.2 | XP_015648280.1 | 4 - 5 | 1166 | 84 | |
| GenBank mRNA | AK103187.1 | - | 4 - 5 | 1130 | 84 | |
More Information
Set Membership: |
|

LOC4345926
Genomic Map