Assay Details
| Gene Symbol: LOC4345926 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792794.2 | XP_015648280.1 | 6 - 7 | 1407 | 80 | |
| GenBank mRNA | AK103187.1 | - | 6 - 7 | 1371 | 80 | |
| AK120031.1 | - | 250 | 80 | |||
More Information
Set Membership: |
|

LOC4345926
Genomic Map