Assay Details
Gene Symbol: LOC4346072 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015794738.2 | XP_015650224.1 | 1 - 2 | 216 | 81 | |
XM_015794739.2 | XP_015650225.1 | 1 - 2 | 227 | 81 | ||
XM_015794740.2 | XP_015650226.1 | 1 - 2 | 227 | 81 | ||
XM_015794741.2 | XP_015650227.1 | 1 - 2 | 227 | 81 | ||
XM_015794742.2 | XP_015650228.1 | 1 - 2 | 186 | 81 | ||
GenBank mRNA | AK070148.1 | - | 1 - 2 | 228 | 81 | |
AK101704.1 | - | 1 - 2 | 218 | 81 | ||
AK105045.1 | - | 1 - 2 | 227 | 81 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015794738.2, XM_015794739.2, XM_015794740.2, XM_015794741.2, XM_015794742.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
