Assay Details
Gene Symbol: LOC4346072 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015794738.2 | XP_015650224.1 | 6 - 7 | 776 | 73 | |
XM_015794739.2 | XP_015650225.1 | 6 - 7 | 787 | 73 | ||
XM_015794740.2 | XP_015650226.1 | 6 - 7 | 784 | 73 | ||
XM_015794741.2 | XP_015650227.1 | 6 - 7 | 787 | 73 | ||
XM_015794742.2 | XP_015650228.1 | 6 - 7 | 743 | 73 | ||
GenBank mRNA | AK070148.1 | - | 6 - 7 | 788 | 73 | |
AK101704.1 | - | 6 - 7 | 778 | 73 | ||
AK105045.1 | - | 6 - 7 | 784 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
