Assay Details
Gene Symbol: LOC4346072 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015794738.2 | XP_015650224.1 | 7 - 8 | 883 | 82 | |
XM_015794739.2 | XP_015650225.1 | 7 - 8 | 894 | 82 | ||
XM_015794740.2 | XP_015650226.1 | 7 - 8 | 891 | 82 | ||
XM_015794741.2 | XP_015650227.1 | 7 - 8 | 894 | 82 | ||
XM_015794742.2 | XP_015650228.1 | 7 - 8 | 850 | 82 | ||
GenBank mRNA | AK070148.1 | - | 7 - 8 | 895 | 82 | |
AK101704.1 | - | 7 - 8 | 885 | 82 | ||
AK105045.1 | - | 7 - 8 | 891 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
