Assay Details
Gene Symbol: LOC4346072 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015794738.2 | XP_015650224.1 | 9 - 10 | 1067 | 131 | |
XM_015794739.2 | XP_015650225.1 | 9 - 10 | 1078 | 131 | ||
XM_015794740.2 | XP_015650226.1 | 9 - 10 | 1075 | 131 | ||
GenBank mRNA | AK101704.1 | - | 9 - 10 | 1069 | 131 | |
AK105045.1 | - | 9 - 10 | 1075 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
