Assay Details
Gene Symbol: LOC4346402 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015756766.2 | XP_015612252.1 | 12 - 13 | 2089 | 86 | |
XM_015756767.2 | XP_015612253.1 | 12 - 13 | 2167 | 86 | ||
GenBank mRNA | AK058665.1 | - | 2 - 3 | 185 | 86 | |
AK067738.1 | - | 11 - 12 | 1448 | 86 | ||
AK100974.1 | - | 12 - 13 | 2090 | 86 | ||
AY224568.1 | - | 10 - 11 | 1261 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
