Assay Details
Gene Symbol: LOC4346565 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795821.2 | XP_015651307.1 | 1 - 2 | 487 | 65 | |
GenBank mRNA | AK121740.1 | - | 1 - 2 | 493 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
