Assay Details
Gene Symbol: LOC4346565 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795821.2 | XP_015651307.1 | 2 - 3 | 600 | 87 | |
GenBank mRNA | AK121740.1 | - | 2 - 3 | 606 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
