Assay Details
Gene Symbol: LOC4346565 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795821.2 | XP_015651307.1 | 5 - 6 | 1037 | 64 | |
GenBank mRNA | AK121740.1 | - | 5 - 6 | 1044 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
