Assay Details
Gene Symbol: LOC4346565 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795821.2 | XP_015651307.1 | 6 - 7 | 1260 | 70 | |
GenBank mRNA | AK121740.1 | - | 6 - 7 | 1267 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
