Assay Details
Gene Symbol: LOC4346565 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795821.2 | XP_015651307.1 | 9 - 10 | 2555 | 96 | |
GenBank mRNA | AK121740.1 | - | 9 - 10 | 2563 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
