Assay Details
| Gene Symbol: LOC4348138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758879.2 | XP_015614365.1 | 3 - 4 | 415 | 116 | |
| GenBank mRNA | AK061152.1 | - | 887 | 116 | ||
| AK121800.1 | - | 3 - 4 | 380 | 116 | ||
More Information
Set Membership: |
|

LOC4348138
Genomic Map