Assay Details
| Gene Symbol: LOC4348636 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759183.2 | XP_015614669.1 | 1 - 2 | 173 | 70 | |
| GenBank mRNA | AB193582.1 | - | 1 - 2 | 34 | 70 | |
| AK069385.1 | - | 1 - 2 | 124 | 70 | ||
| AK104822.1 | - | 1 - 2 | 119 | 70 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015759183.2 transcripts.
Set Membership: |
|

LOC4348636
Genomic Map