Assay Details
| Gene Symbol: LOC4348636 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759183.2 | XP_015614669.1 | 6 - 7 | 1152 | 61 | |
| GenBank mRNA | AB193582.1 | - | 6 - 7 | 1013 | 61 | |
| AK069385.1 | - | 6 - 7 | 1103 | 61 | ||
| AK104822.1 | - | 6 - 7 | 1098 | 61 | ||
More Information
Set Membership: |
|

LOC4348636
Genomic Map