Assay Details
| Gene Symbol: LOC4349116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757235.2 | XP_015612721.1 | 7 - 8 | 884 | 69 | |
| GenBank mRNA | AK065934.1 | - | 8 - 9 | 686 | 69 | |
More Information
Set Membership: |
|

LOC4349116
Genomic Map