Assay Details
Gene Symbol: LOC4349412 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015758782.2 | XP_015614268.1 | 6 - 7 | 709 | 144 | |
GenBank mRNA | AK243065.1 | - | 6 - 7 | 712 | 144 | |
DQ201195.1 | - | 6 - 7 | 464 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
