Assay Details
Gene Symbol: LOC4349698 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015761665.2 | XP_015617151.1 | 12 - 13 | 1417 | 110 | |
XM_015761666.2 | XP_015617152.1 | 12 - 13 | 1403 | 110 | ||
GenBank mRNA | AK112043.1 | - | 11 - 12 | 1005 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
