Assay Details
Gene Symbol: LOC4349698 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015761665.2 | XP_015617151.1 | 13 - 14 | 1475 | 126 | |
XM_015761666.2 | XP_015617152.1 | 13 - 14 | 1461 | 126 | ||
GenBank mRNA | AK112043.1 | - | 12 - 13 | 1063 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
