Assay Details
| Gene Symbol: LOC107277152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763381.2 | XP_015618867.1 | 1 - 2 | 241 | 98 | |
| Gene Symbol: LOC9270331 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761784.2 | XP_015617270.1 | 1 - 2 | 255 | 98 | |
More Information
Set Membership: |
|

LOC107277152
Genomic Map