Assay Details
| Gene Symbol: LOC9272231 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015760032.2 | XP_015615518.1 | 11 - 12 | 1591 | 90 | |
| GenBank mRNA | AK241662.1 | - | 10 - 11 | 2271 | 90 | |
More Information
Set Membership: |
|

LOC9272231
Genomic Map