Assay Details
| Gene Symbol: LOC4351922 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764056.1 | XP_015619542.1 | 2 - 3 | 264 | 73 | |
| GenBank mRNA | AK069945.1 | - | 2 - 3 | 261 | 73 | |
More Information
Set Membership: |
|

LOC4351922
Genomic Map