Assay Details
| Gene Symbol: LOC4351922 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764056.1 | XP_015619542.1 | 7 - 8 | 819 | 111 | |
| GenBank mRNA | AK069945.1 | - | 7 - 8 | 816 | 111 | |
More Information
Set Membership: |
|

LOC4351922
Genomic Map