Assay Details
Gene Symbol: LOC9269535 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015764715.2 | XP_015620201.1 | 13 | 3847 | 134 | |
XM_015764716.2 | XP_015620202.1 | 13 | 3893 | 134 | ||
GenBank mRNA | AK059486.1 | - | 1 | 1017 | 134 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
