Assay Details
Gene Symbol: LOC4350220 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015762156.2 | XP_015617642.1 | 1 - 2 | 287 | 63 | |
GenBank mRNA | AK107906.1 | - | 1 - 2 | 257 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
