Assay Details
Gene Symbol: LOC4351224 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015761411.1 | XP_015616897.1 | 1 - 2 | 165 | 136 | |
GenBank mRNA | AB034698.1 | - | 1 - 2 | 101 | 136 | |
AK060847.1 | - | 1 - 2 | 102 | 136 | ||
AK064986.1 | - | 1 - 2 | 103 | 136 | ||
AK104332.1 | - | 1 - 2 | 102 | 136 | ||
AK119513.1 | - | 1 - 2 | 114 | 136 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015761411.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
