Assay Details
Gene Symbol: LOC4351224 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015761411.1 | XP_015616897.1 | 4 - 5 | 1124 | 70 | |
GenBank mRNA | AB034698.1 | - | 4 - 5 | 1060 | 70 | |
AB110180.1 | - | 4 - 5 | 1025 | 70 | ||
AK060841.1 | - | 3 - 4 | 845 | 70 | ||
AK060847.1 | - | 4 - 5 | 1061 | 70 | ||
AK064986.1 | - | 4 - 5 | 1062 | 70 | ||
AK104332.1 | - | 5 - 6 | 1024 | 70 | ||
AK119513.1 | - | 4 - 5 | 1073 | 70 | ||
HQ324826.1 | - | 4 - 5 | 1041 | 70 | ||
KC140121.1 | - | 4 - 5 | 1033 | 70 | ||
U74321.1 | - | 4 - 5 | 1030 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
