Assay Details
Gene Symbol: LOC4351224 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK060841.1 | - | 4 - 5 | 1132 | 74 | |
AK060847.1 | - | 5 - 6 | 1348 | 74 | ||
AK064986.1 | - | 5 - 6 | 1349 | 74 | ||
AK104332.1 | - | 6 - 7 | 1311 | 74 | ||
AK119513.1 | - | 5 - 6 | 1360 | 74 | ||
HQ324826.1 | - | 5 - 6 | 1328 | 74 | ||
KC140121.1 | - | 5 - 6 | 1320 | 74 | ||
U74321.1 | - | 5 - 6 | 1317 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
