Assay Details
Gene Symbol: LOC4351224 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK060841.1 | - | 5 - 6 | 1210 | 64 | |
AK060847.1 | - | 6 - 7 | 1426 | 64 | ||
AK064986.1 | - | 6 - 7 | 1427 | 64 | ||
AK104332.1 | - | 7 - 8 | 1389 | 64 | ||
AK119513.1 | - | 6 - 7 | 1438 | 64 | ||
HQ324826.1 | - | 6 - 7 | 1406 | 64 | ||
KC140121.1 | - | 6 - 7 | 1398 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
